Learn More
Enzo Life Sciences E6 Berbamine (10mg). CAS: 114784-59-7
Supplier: Enzo Life Sciences BMLCA3020010
A potent, selective, cell-permeable calmodulin antagonist. It inhibits calmodulin-dependent enzymes such as MLC kinase (IC50=8 µM) and phosphodiesterase I (IC50=0.53 µM). Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.