Learn More
Medchemexpress LLC HY-107456 10mg Medchemexpress, E6130 CAS:1427058-33-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10745610MG
Encompass_Preferred
Medchemexpress, HY-107456 10mg E6130 CAS:1427058-33-0 E6130 is an orally available and highly selective CX3CR1 modulator, that may be effective for treatment of inflammatory bowel disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8419