Learn More
Selleck Chemical LLC E6446 S0716-100MG
SDP
Supplier:  Selleck Chemical LLC S0716100MG
Encompass_Preferred
E6446 is an inhibitor of Toll-like receptor (TLR) 7 and 9 signaling in a variety of human and mouse cell types and inhibits DNA-TLR9 interaction in vitro
Catalog No. 50-000-42895