Learn More
TARGETMOL CHEMICALS INC E6446 DIHYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T40685MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. E6446 dihydrochloride is a novel synthetic antagonist for nucleic acid-sensing TLRs; potently suppressed DNA stimulation of HEKTLR9 cells with IC50 of 10 nM. purity: 99%
Catalog No. 50-257-9975