Learn More
TARGETMOL CHEMICALS INC E7820 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T443525MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. E7820 (ER68203-00) is an angiogenesis inhibitor by suppressing integrin a2 (a cell adhesion molecule expressed on endothelial cells). purity: 99%
Catalog No. 50-257-9884