Learn More
Abcam E7820, 25MG
Supplier:  Abcam AB28710625MG
Encompass_Preferred
MW 336.4 Da, Purity >=98%. A small molecule sulfonamide derivative with potential antiangiogenic and antitumor activities. E7820 inhibits angiogenesis by suppressing integrin alpha 2, a cell adhesion molecule expressed on endothelial cells. Inhibition of integrin alpha 2 leads to an inhibition of cell-cell interactions, endothelial cell-matrix interactions, vascular endothelial cell proliferation and angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-0459