Learn More
Cayman Chemical EA4 Fatty Acids and Derivative
SDP
Supplier:  Cayman Chemical 101115 MG
Encompass_Preferred
An inhibitor of PLA2 from red blood cells (Ki = 130 µM); inhibits the ionophore-induced arachidonic acid release from human and bovine RBCs; induces mRNA expression and increases protein levels of CYP1A1 in murine hepatoma Hepa-1c1c7 cells; modulates CYP1A1 and CYP1B1 expression in MCF-7, Hep-G2, and HL-60 cells
Catalog No. 50-288-7839