Learn More
Medchemexpress LLC EAAT2 activator 1 | 892415-28-0 | 98.91% | 331.80 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY139692100MG
Encompass_Preferred
EAAT2 activator 1 is a potent activator of excitatory amino acid transporter 2 (EAAT2), which is the primary glutamate transporter responsible for removing glutamate from synapses. It has been shown to increase EAAT2 protein levels in a dose-dependent manner.
  • Potent activator of excitatory amino acid transporter 2 (EAAT2)
  • Increases EAAT2 protein levels dose-dependently
  • Solid form
  • Light brown to brown color
Catalog No. 50-003-50139