Learn More
Medchemexpress LLC Eact | 461000-66-8 | 98.9% | 428.50 | C22H24N2O5S | 5 MG

Supplier: Medchemexpress LLC HY1033685MG
Eact is a small-molecule activator of the calcium-activated chloride channel TMEM16A (ANO1) that also directly activates TRPV1 channels in sensory nociceptors. It is used as a pharmacological tool to study TMEM16A-mediated chloride currents and TRPV1-dependent nociception and itch responses in vitro and in vivo.
- Selective activator of TMEM16A and activator of TRPV1.
- Molecular formula C22H24N2O5S; molecular weight 428.50.
- Purity approximately 98.9%, suitable for biochemical and cellular assays.
- Soluble in DMSO (50 mg/mL); in vivo formulation guidance: 10% DMSO/90% corn oil, solubility ≥ 2.5 mg/mL.
- Powder storage: -20°C (3 years) or 4°C (2 years); solvent storage: -80°C (2 years) or -20°C (1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.