Learn More
New England Biolabs, Inc. EaeI – 1000 units
SDP
Supplier:  New England Biolabs, Inc. R0508L
Encompass
A restriction endonuclease that recognizes the sequence Y^GGCC_R.
Catalog No. 50-812-153