Learn More
ATCC Eagle's Minimum Essential Medium (EMEM), 500mL
Supplier:  ATCC 302003/DEL
Encompass
It is modified to contain Earles Balanced Salt Solution, non-essential amino acids, 2 mM L-glutamine, 1 mM sodium pyruvate, and 1500 mg/L sodium bicarbonate. This reduced level of sodium bicarbonate (NaHCO3, 1.5 g/L) is intended for use in 5% CO2 in air. Additional sodium bicarbonate may be required for use in incubators containing higher percentages of CO2. For use by FOR-PROFIT companies only; NON-PROFIT companies use 50188268NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-188-268FP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product