Learn More
MSC 3M Earplugs, 33 dB NRR, Foam, Cylinder, Corded
Supplier:  MSC 85305464
Encompass
These earplugs are designed to provide hearing protection in noisy environments. They feature a 33 dB Noise Reduction Rating, a corded design for convenience, and a comfortable cylindrical shape made from foam. Ideal for various industrial and personal applications, they are reusable and come in a high-visibility color.
  • 33 dB Noise Reduction Rating for effective hearing protection
  • Foam material (PVC) for comfort and secure fit
  • Cylinder shape for easy insertion and fit
  • Corded design with vinyl cord to prevent loss
  • Reusable for extended use
  • Non-metal detectable
  • High-visibility yellow plug with a blue cord
  • Large size to fit a variety of users
  • Packaged with 200 pairs per box
Catalog No. 50-283-5498