Learn More
Essendant EARPLUGSPELTORMUFF
Supplier:  Essendant MMMH7A
Encompass_Preferred
Deluxe muffs stand up to high-noise environments- airport runways construction sites weapons-testing fields etc Liquid- and foam-filled ear cups significantly attenuate both low and high frequencies Padded headband adjusts to provide a slop-free fit Assembly is color-coded for compliance sighting Material(s)- Metal Plastic Ear Cushion Material- Liquid/Foam Noise Reduction Rate (NRR)- 27 dB Ear Muff Style- Over-The-Head
Catalog No. 50-000-35848