Learn More
Essendant EARPLUGSRSOFT BLSTSNEYL
Supplier:  Essendant MMM3111252
Encompass_Preferred
These foam earplugs feature soft slow-recovery foam for comfort and outstanding noise reduction A red flame design adds fun to serious hearing protection Soft foam earplugs are easy to roll down and insert then expand within the earcanal for a comfortable secure fit Poly bag keeps each pair of earplugs clean before use Regular size fits most earcanals Test compatible with 3M E-A-Rfit Validation System CSA Class AL Research suggests that many users will receive less noise reduction than indicated by the NRR due to variation in earplug fit earplug fitting skill and motivation of the user It is recommended that the NRR be reduced by 50pct to better estimate typical workplace protection Corded/Cordless Pair- Corded Shape- Tapered Material(s)- Polyurethane Noise Reduction Rate (NRR)- 33 dB
Catalog No. 50-000-31200