Learn More
Essendant EARPLUGSULTRAFTCSE/CORD
Supplier:  Essendant MMM3404002
Encompass_Preferred
Choose E-A-R UltraFit Earplugs if you re looking for an excellent hearing-protection option that provides improved hygiene performance and a wonderfully comfortable wearing experience Triple-flange design contours itself to a wide variety of ear-canal shapes and sizes to ensure a more effective noise-blocking seal making them more practical and more versatile too Almost anyone can find a snug comfortable fit with a pair of UtraFit Flexi-fit stem and no-roll tips allow the wearer to insert the plugs without touching the earplugs tips keeping the tips and the user s ears clean even when the user s fingers or gloves aren t All UltraFit Earplugs are compatible with the 3M E-A-Rfit Dual-Ear Validation System E-A-Rfit provides an accurate quantitative assessment of exactly how much noise-reduction a hearing protector provides to any given worker Triple-flange tips are washable and reusable (handwash with soap and water only) Blue Cords/Carry Cases Corded/Cordless Pair- Corded
Catalog No. 50-000-31199