Learn More
Agilent Technologies EasiVial PS-M, 2 mL, 30 vials/pack, 4 standards/vial, GPC/SEC calibration, molecular weight range 162-400,000 g/mol
Supplier:  Agilent Technologies PL20100301
Encompass
The Agilent EasiVial PS-M is a pre-weighed calibration kit designed to enhance the accuracy and efficiency of Gel Permeation Chromatography and Size Exclusion Chromatography column calibrations. This kit features 30 polystyrene standards housed in 2 mL vials, yielding 12 calibration points from just three injections. By automating the weighing process, it not only increases precision but also decreases the risks involved with solvent handling, making it an ideal solution for diverse GPC applications.
  • Pre-weighed standards improve calibration accuracy.
  • Rapid calibration process saves time.
  • Minimized solvent handling reduces risks.
  • Applicable for various GPC/SEC setups.
  • Color-coded vials ensure easy identification.
Catalog No. NC2069178