Learn More
Agilent Technologies Includes 10 each of 3 different vials containing pre-weighed standards with 4 different molecular masses, total 12 molecular masses, nominal Mp 162 - 7,000,000 Da, 3 x 10 vials, 2 mL
Supplier:  Agilent Technologies PL20100201
Encompass
EasiVial PS-H, 2 mL Polystyrene calibration kit. Includes 10 each of 3 different vials containing pre-weighed standards with 4 different molecular masses, total 12 molecular masses, nominal Mp 162 - 7,000,000 Da, 3 x 10 vials, 2 mL
Catalog No. NC2949810