Learn More
Alpco Ltd EASY STOOL EXTRACTION DEVICE
SDP
Supplier:  Alpco Ltd 30EZEX100
Encompass
The Easy Stool Extraction Device is an FDA Class I Exempt tool that can be used across four comprehensive gastroenterology ELISA testing panels comprised of 13 different analytes.
The device allows lab technicians to simplify stool sample processing and replace manual weighing with one simple step. A grooved plastic wand is dipped into a stool sample in multiple areas and placed inside a vial pre-filled with a universal extraction buffer. This standardized device accurately measures 15 mg of stool and extracts the sample in 1.5 mL of extraction buffer, yielding a final dilution of 1:100. The resulting extract can be placed directly on automated platforms and run across up to 13 different assays including:
Catalog No. NC2283521