Learn More
XenoTech,LLC EASYCYP CYP4F3BLR+ 0.5 NMOL
SDP
Supplier:  XenoTech,LLC CYP/EZ057
Encompass
EasyCYP CYP4F3B Low Reductase, plus b5, 0.5 nmol P450 at 1 nmol/ml, 10 mg/ml protein
Catalog No. 50-142-943