Learn More
STEMCELL Technologies EasyPlate™ EasySep™ Magnet, For processing: 1 Unit
Supplier: STEMCELL Technologies 18102
The EasyPlate™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a small number of cells (up to 2 x 10^7 cells per well). The EasyPlate™ EasySep™ Magnet generates a high-gradient magnetic field that is strong enough to separate cells labeled with EasySep™ D Magnetic Particles without the use of columns. The EasyPlate™ EasySep™ Magnet is designed to hold a round-bottom non-tissue culture-treated 96-well plate.