Learn More
STEMCELL Technologies EasySep Direct Human CTC Enrichment Kit, For processing: 100 mL whole blood processed
SDP
Supplier:  STEMCELL Technologies 19657
Encompass
The EasySep Direct Human CTC Enrichment Kit can be used to enrich Circulating Tumor Cells (CTCs) directly from human whole blood by immunomagnetic negative selection without the need for either density gradient centrifugation or RBC lysis. Isolated cells are immediately ready for downstream analysis or assays.
Catalog No. 50-239-0118