Learn More
STEMCELL Technologies EasySep™ Direct Human Eosinophil Isolation Kit, For processing: 100 mL whole blood processed
SDP
Supplier:  STEMCELL Technologies 19656
Encompass
The EasySep™ Direct Human Eosinophil Isolation Kit isolates functional, highly purified eosinophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation, or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.
Catalog No. NC1736329