Learn More
STEMCELL Technologies EasySep™ EasyStand™ x 4 with Magnets, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 18135
Encompass
The EasySep™ EasyStand™ facilitates the separation of one or more samples at one time. Each EasySep™ EasyStand™ can hold a single EasySep™ Magnet (Catalog #18000), and a simple system links up to 6 individual EasySep™ EasyStands™ together. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes.
Catalog No. 50-197-4631
Only null left
Add to Cart
Request a Quote