Learn More
STEMCELL Technologies EasySep™ Human Pan-ILC Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 17975
Encompass
The EasySep™ Human Pan-ILC Enrichment Kit is designed to enrich Group 1, 2, and 3 Innate Lymphoid Cells (ILC1, 2, and 3) from leukapheresis products by immunomagnetic negative selection. Unwanted cells are targeted for removal by Tetrameric Antibody Complexes directed against non-ILCs and dextran-coated magnetic particles (RapidSpheres™). Labeled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube as enriched ILCs are simply poured off into a new tube. Isolated cells are immediately available for flow sorting and other downstream applications.
Catalog No. NC1395719