Learn More
STEMCELL Technologies EasySep™ Human ILC2 Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 17972
Encompass
The EasySep™ Human ILC2 enrichment kit is designed to enrich Group 2 Innate Lymphoid Cells (ILC2s) from leukapheresis products by immunomagnetic negative selection. Unwanted cells are targeted for removal by Tetrameric Antibody Complexes directed against non-ILC2s and dextran-coated magnetic particles (RapidSpheres™). Labeled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube as desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry and cell sorting.
Catalog No. NC1667322
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov