Learn More
STEMCELL Technologies EasySep™ Human Memory CD8+ T Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 19159
Encompass
The EasySep™ Human Memory CD8+ T Cell Enrichment Kit is designed to isolate memory CD8+ T cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD4, CD14, CD16, CD19, CD20, CD34, CD36, CD45RA, CD56, CD61, CD66b, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. NC0887384
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov