Learn More
STEMCELL Technologies EasySep™ Human PSC-Derived Cardiomyocyte Enrichment Kit, For processing: 2.5 x 10^8 cells
SDP
Supplier:  STEMCELL Technologies 17965
Encompass
The EasySep™ Human PSC-Derived Cardiomyocyte Enrichment Kit is designed to isolate human PSC-derived cardiomyocytes from cell cultures by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-cardiomyocytes and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. 50-197-4621