Learn More
STEMCELL Technologies EasySep™ Multistand with "The Big Easy" EasySep™ Magnets, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 18100
Encompass
The EasySep™ Multistand allows the separation of up to 4 samples at one time. An ingenious reversible clamping bar allows the Multistand to hold up to 4 EasySep™ (Catalog #18000) or 4 "The Big Easy" EasySep™ Magnets (Catalog #18001). After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes. The receiving tube bar is indexed to ensure proper position of the tubes.
Catalog No. 50-197-4628
Only null left
Add to Cart
Request a Quote