Learn More
STEMCELL Technologies EasySep™ Release Human CD4 Positive Selection Kit, For processing: 1 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 17752
Encompass
The EasySep™ Release Human CD4 Positive Selection Kit is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection.Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated CD4+ cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands. The CD4 antigen is strongly expressed on helper T cells and is expressed more weakly on peripheral blood monocytes and tissue macrophages.
Catalog No. NC1342621
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov