Learn More
Sigma Aldrich Fine Chemicals Biosciences Eatons Reagent
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 380814100ML
Encompass
Eaton Reagent contains 7.7wt% phosphorus pentoxide in methanesulfonic acid. It is commonly used as a catalyst and condensing agent.
Catalog No. 50-289-8548