Learn More
Electron Microscopy Sciences MAC Electron Backscatter Particle Analysis Standard
SDP
Supplier:  Electron Microscopy Sciences 80070EBP
Encompass_Preferred
In 13 mm Aluminium pin stub certified standards for system calibration. Diamond polished to 0.25um surface finish. Carbon Coated. This Electron Backscatter Particle Analysis standard allows user to go across all grey areas.
Catalog No. 50-190-1614