Learn More
Medchemexpress LLC Ebaresdax | 1334471-39-4 | 99.9% | 266.32 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY139569100MG
Encompass_Preferred
Ebaresdax (ACP-044) is a chemical compound that inhibits peroxynitrite oxidation and peroxynitrite mediated cytotoxicity. It is a white to off-white solid, classified as an alkaloid and phenol, with an initial source as an endogenous metabolite. It has been involved in clinical trials for acute postoperative pain and osteoarthritis pain. It is for research use only.
  • Inhibits peroxynitrite oxidation
  • Inhibits peroxynitrite mediated cytotoxicity
  • Appears as a white to off-white solid
  • Classified as an alkaloid and a phenol
  • Initial source is an endogenous metabolite
  • Involved in clinical trials for acute postoperative pain
  • Involved in clinical trials for osteoarthritis pain
  • For research use only
Catalog No. 50-003-50994