Learn More
Medchemexpress LLC Ebaresdax | 1334471-39-4 | 99.9% | C12H14N2O3S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13956910MG
Encompass_Preferred
Ebaresdax (ACP-044) is an investigational analgesic research compound reported to inhibit SIN-1-derived peroxynitrite oxidation and peroxynitrite-mediated cytotoxicity. It is provided for research use only and is intended for in vitro studies of peroxynitrite-related signaling and cytotoxic mechanisms.
  • Displays potent inhibition of peroxynitrite oxidation (IC50 = 3.7 ± 0.80 μM).
  • Reduces peroxynitrite-mediated cytotoxicity (IC50 = 0.13 ± 0.02 μM).
  • High purity and well characterized for research applications.
  • Available in solid and solution formats for flexible assay setup.
  • Supplied with supporting analytical documents (datasheet, COA, SDS).
Catalog No. 50-002-60876