Learn More
Medchemexpress LLC HY-111418 10mg Medchemexpress, EBI-2511 CAS:2098546-05-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11141810MG
Encompass_Preferred
Medchemexpress, HY-111418 10mg EBI-2511 CAS:2098546-05-3 EBI-2511 is a highly potent and orally active EZH2 inhibitor, with an IC50 of 6 nM in Pfeffiera cell lines, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8863