Learn More
Genscript Corporation eBlot L1
Supplier:  Genscript Corporation L00686
Encompass
eBlot L1 is a highly efficient wet protein transfer system which uses a patented technology developed by GenScript eBlot L1 combines the high stability and efficiency of the traditional wet transfer with the rapidness and convenience of the semi-dry transfer system The device allows the efficient transfer of small medium as well as large molecular weight proteins within 15 minutes
Catalog No. 50-000-58419