Learn More
Cayman Chemical Ebola VIrus VP24subTYP ZaI
SDP
Supplier:  Cayman Chemical 41067100 UG
Encompass_Preferred
Source: Recombinant Ebola virus N-terminal His-tagged VP24 expressed in E. coli • Amino acids: 1-233 • MW: 28.5 kDa
Catalog No. 50-294-4918