Learn More
Medchemexpress LLC Ebola virus VP40/Mat 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP768915UG
Encompass_Preferred
Ebola virus VP40 (matrix VP40 protein) centrally directs virus assembly and budding and has complex interactions with viral ribonucleocapsid and host ESCRT proteins (VPS4 PDCD6IP/ALIX NEDD4 TGS101) Ebola virus VP40/Matrix VP40 Protein (AHX24648 His) is the recombinant Virus-derived Ebola virus VP40/Matrix VP40 protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-85918