Learn More
Medchemexpress LLC HY-137498 5mg Medchemexpress, EBOV/MARV-IN-1 CAS:2479465-67-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1374985MG
Encompass_Preferred
Medchemexpress, HY-137498 5mg EBOV/MARV-IN-1 CAS:2479465-67-1 EBOV/MARV-IN-1 is a potent inhibitor of Ebola virus (EBOV) and Marburg virus (MARV), with broad-spectrum activity (EC50=0.31, and 0.82 µM, respectively) and low cytotoxicity (SI>100) in HeLa cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1495