Learn More
Medchemexpress LLC EBP-IN-1 | 3034437-44-7 | 99.8% | 411.54 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1623515MG
Encompass_Preferred
EBP-IN-1 (compound 11) is an inhibitor of emopamil-binding protein (EBP), a sterol isomerase in the cholesterol biosynthetic pathway. It has a long half-life in rodents and possesses good metabolic turnover and brain penetration properties. EBP-IN-1 enhances oligodendrocyte formation in human cortical organoids. It is intended for research use only.
  • Inhibitor of emopamil-binding protein (EBP)
  • Long half-life in rodents
  • Good metabolic turnover
  • Brain penetration properties
  • Enhances oligodendrocyte formation in human cortical organoids
Catalog No. 50-003-94533