Learn More
Medchemexpress LLC HY-13750 100mg , Ebselen CAS:60940-34-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13750100MG
Encompass_Preferred
HY-13750 100mg Ebselen CAS: 60940-34-3 Ebselen (SPI-1005), a glutathione peroxidase mimetic, is a potent voltage-dependent calcium channel (VDCC) blocker. Ebselen potently inhibits Mpro (IC50=0.67 μM) and COVID-19 virus (EC50=4.67 μM).Ebselen is an inhibitor of HIV-1 capsid CTD dimerization. Ebselen, an organoselenium compound, can permeate the blood-brain barrier and has anti-inflammatory, antioxidant and anticancer activity[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0100