Learn More
Enzo Life Sciences Ebselen (25mg). CAS: 60940-34-3
SDP
Supplier:  Enzo Life Sciences ALX270097M025
Encompass_Preferred
Organoselenium compound. Antioxidant. Selenium-based inhibitor of protein kinase C, NADPH, 5-lipoxygenase, cyclooxygenase (COX) and NADPH oxidase. Anti-inflammatory antioxidant. Mimics glutathione peroxidase. Inhibits oxidative modifications of low density lipoproteins (LDL). Protects cerebellar granule neurons against 4-hydroxynonenal-induced neuronal death. Formula: C13H9NOSe. MW: 274.2. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8404