Learn More
AdipoGen Ebselen (25 mg)
SDP
Supplier:  AdipoGen AGCR10031M025
Encompass_Preferred
Chemical. CAS: 60940-34-3. Formula: C13H9NOSe. MW: 274.2. Glutathione peroxidase mimetic. Peroxynitrite scavenger. Anti-inflammatory Antioxidant. Protein kinase C (PKC), NADPH, lipoxygenase, COX, NOS, H+-ATPase and NADPH oxidase inhibitor. Antifungal. Review. Potent IDO-1 inhibitor with Ki 94nM. Inhibitor of HIV-1 capsid C-terminal domain dimerization.
Catalog No. 50-205-3783