Learn More
Abcam Ebselen, mimic of glutathione pero x idase, 5MG
Supplier:  Abcam AB1424245MG
Encompass_Preferred
MW 274.19 Da, Purity >98%. Seleno-organic compound, antioxidant, mimic of glutathione peroxidase. Inhibitor of lipoxygenases, NO synthases, NADPH oxidase, protein kinase C and H+/K(+)-ATPase producing anti-inflammatory effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6246