Learn More
Medchemexpress LLC Ebselen oxide 1mg | 104473-83-8 | 290.18 g/mol | C13H9NO2Se | 1 MG

Supplier: Medchemexpress LLC HY1145481MG
Ebselen oxide is the selenone analogue of ebselen supplied as a research-grade biochemical reagent. It functions as a diguanylate cyclase (DGC) inhibitor by covalently modifying target enzymes and is used in enzyme inhibition and related biochemical studies.
- Used as a biochemical research reagent and DGC inhibitor.
- Covalently modifies diguanylate cyclase to inhibit c-di-GMP-receptor interactions.
- Purity ≥98.0% as supplied for research use.
- Molecular formula C13H9NO2Se and molecular weight 290.18 g/mol.
- Supplied in small milligram quantities suitable for laboratory assays.
- Commonly applied in enzyme inhibition and microbial signaling research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.