Learn More
Cayman Chemical Ebselen Oxide Fatty Acids and
SDP
Supplier:  Cayman Chemical 1001229825 MG
Encompass_Preferred
Formed by the oxidation of ebselen; lacks antioxidant acitivity
Catalog No. 50-288-9054