Learn More
Medchemexpress LLC Ebsulfur | 2527-03-9 | 99.4% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12817010MG
Encompass_Preferred
Ebsulfur is a benzisothiazolone cytocidal inhibitor designed to target the trypanothione reductase (TryR) of Trypanosoma brucei. It is a valuable compound for research focused on African trypanosomiasis.
  • Targets trypanothione reductase (TryR) of Trypanosoma brucei
  • IC50 values of 61-293 nM
  • Useful for research on African trypanosomiasis
Catalog No. 50-003-40904