Learn More
Medchemexpress LLC EBV BZLF1 (190-197) | 200124-17-0 | 1003.24 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP10497A
Encompass_Preferred
EBV BZLF1 (190-197) is an immunodominant CEF-controlling peptide derived from Epstein-Barr virus (EBV). It is a suitable epitope for CD8+ T cells to stimulate the release of IFNg. CTL responses to EBV BZLF1 (190-197) are diverse and cross-recognize similar peptides from both host self-proteins and bacterial proteins. It can be used to study autoimmune diseases.
  • Immunodominant CEF-controlling peptide from Epstein-Barr virus (EBV)
  • Suitable epitope for CD8+ T cells to stimulate IFNg release
  • CTL responses are diverse and cross-recognize similar peptides
  • Useful for studying autoimmune diseases
Catalog No. 50-004-26578