Learn More
Medchemexpress LLC EBV EBNA3B (416-424) | 147138-29-2 | 99.52% | 1021.21 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP106645MG
Encompass_Preferred
EBV EBNA3B (416-424) is an HLA-A11-restricted peptide antigen derived from EBV EBNA3B protein.
Catalog No. 50-004-25554