Learn More
Eenzyme LLC EBV-HHV-4-ENVELOP GLYCOPR 50UG
Supplier:  Eenzyme LLC HHV4GP42P
Encompass
EBV (HHV-4) Envelope Glycoprotein 42 50ug. Name: Epstein–Barr virus envelope glycoprotein 42; Purity: >95% (by 10% SDS-PAGE gel); Source: Expressed and purified from HEK293 cells
Catalog No. NC2379414