Learn More
Santa Cruz Biotechnology EBV ZEBRA Antibody (BZ1) | 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC53904
Encompass
Anti-EBV ZEBRA Antibody (BZ1) is a mouse monoclonal IgG1 κ EBV ZEBRA antibody, cited in 95 publications, provided at 200 µg/ml. Raised against full-length recombinant EBV ZEBRA protein. EBV ZEBRA Antibody (BZ1) is recommended for detection of ZEBRA of EBV origin by WB, IP, IF and ELISA.
Catalog No. NC1875642